Learn More
Quickfit™ Reduction Socket Adapter
Product Code. 10203720
Change view
Click to view available options
Cone Size:
Cone Size: 14/23
Cone Size: 19/26
Cone Size: 24/29
Cone Size: 29/32
Cone Size: 34/35
Cone Size: 40/38
Cone Size: 45/40
Cone Size: 50/42
Cone Size: 55/44
Standard Taper Joint No.:
10/19
14/23
19/26
24/29
29/32
34/35
Unit Size:
Each
Pack of 2
Pack of 5
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size Standard Taper Joint No. unitSize
10203720 Cone Size: 19/26 14/23 Pack of 2
10520681 Cone Size: 24/29 14/23 Pack of 2
12634891 Cone Size: 24/29 19/26 Pack of 2
12418876 Cone Size: 29/32 14/23 Pack of 2
12520337 Cone Size: 14/23 10/19 Pack of 2
10765861 Cone Size: 29/32 19/26 Pack of 2
12448876 Cone Size: 34/35 19/26 Pack of 2
12989253 Cone Size: 34/35 24/29 Pack of 2
12654891 Cone Size: 29/32 24/29 Pack of 2
12478876 Cone Size: 34/35 29/32 Pack of 2
12050103 Cone Size: 50/42 34/35 Each
15212806 Cone Size: 45/40 34/35 Each
15597790 Cone Size: 40/38 24/29 Pack of 5
15507800 Cone Size: 45/40 24/29 Pack of 5
15517800 Cone Size: 50/42 24/29 Pack of 5
15527800 Cone Size: 55/44 24/29 Pack of 5
15537800 Cone Size: 45/40 29/32 Pack of 5
15547800 Cone Size: 40/38 34/35 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Product Code. 10203720 Supplier Quickfit Supplier No. DA12
Only null left
Add to basket
Add to basket
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
  • Useful for the connection of dissimilar sized ground glass joints
  • Small sized socket at the top, large sized cone at the bottom
TRUSTED_SUSTAINABILITY
Specifications
Cone Size Cone Size: 19/26
For Use With (Equipment) Dissimilar sized ground Glass joint
Includes Small socket, large cone
Material Borosilicate Glass
Product Type Reduction adapter
Standard Taper Joint No. 14/23