Learn More
Quickfit™ Receiver with Vented Bend
Product Code. p-8003005
Click to view available options
Height (Metric):
80 mm
106 mm
120 mm
Cone Size:
14/23
19/26
24/29
Unit Size:
Each
Product Code. 11342095
Brand: Quickfit RA2/22
Only null left
Add to basket
Add to basket
Receive distillate in a round bottomed flask. The QuickFit™ reciever is jointed with ventend bend to avoid the problems of closed systems.
Receive distillate in a round bottomed flask. The QuickFit™ reciever is jointed with ventend bend to avoid the problems of closed systems.
  • Bend with vent
  • With sidearm to provide vent to atmosphere
  • Avoids the problems of closed systems
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Height (Metric) 106 mm
Height (English) 4.17 in.
Includes Side arm
Cone Size 19/26
Product Type Receiver Adapter