Learn More
Quickfit™ PTFE Screwthread Washers
Product Code. p-8003003
Click to view available options
Thread Size:
13
18
24
28
Diameter (Metric):
10.5 to 11.5 mm (through cap)
6 to 7 mm (through cap)
7 to 8.5 mm (through cap)
8.5 to 9.5 mm (through cap)
Unit Size:
Pack of 10
Product Code. 12939283
Brand: Quickfit QW13/6
Secure screwcaps to glass screwthreads. QuickFit™ screwthread washers for QuickFit™ screwthreads ensure that the cap fits correctly. Washers come in a variety of sizes.
  • For use with Quickfit™ glass screwthreads
  • Melamine screwcaps with aperture at the top
  • Silicone rubber rings with PTFE washers
  • To determine what components are required: i) Check the size of the glass screw thread (size designation refers to O. D. of thread inmm) ii) Check the O. D. of the sampling tube, thermometer, probe etc. , that is to be passed through the screw cap.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 6 to 7 mm (through cap)
Material PTFE
For Use With For Quickfit™ Glass screw threads
Thread Size 13
Product Type Washer