Learn More
Quickfit™ Polyacetal Clamp for Spherical Joints
For retaining glassware having Quickfit™ spherical joints
Brand: Quickfit KS29
517.75 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.06000kg
Description
- Manufactured from polyacetal
- Can considerably reduce the need for clamping
- Easy to fit and remove
- Can be used at temperatures up to 80°C
- Clips are color coded for easy identification
Specifications
5/Pk. | |
Joint Clamp | |
Polyacetal | |
Dark Red |