Learn More
Quickfit™ Plain Short Stem Adapter
Feed distillate from condensers into receiving vessels. QuickFit™ plain short stem adapters come in a wide variety of lengths from short stems to very long ones.
Brand:  Quickfit RA1/01
323.95 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 12425366
  • 341.00 NOK / Each
Estimated Shipment: 12-06-2024
Description
Feed distillate from condensers into receiving vessels. QuickFit™ plain short stem adapters come in a wide variety of lengths from short stems to very long ones.
  • With short or long stem
  • Useful for feeding distillate from condensers into receiving vessels
Specifications
Specifications
Receiver Adapter
2.55 in.
Feeding distillate from condensers into receiving vessels
Borosilicate Glass
65 mm
14/23