Learn More
Quickfit™ Metal Clamps for Flat Flange Joints
Description
Offered in two versions: fork design and spring wire design. Quickfit™ Metal Clamps for Flat Flange Joints are designed to secure corresponding Quickfit™ glassware with flat flange joints.
Two distinct styles to meet joint needs
- Fork design: spring loaded to ensure balanced positive pressure to both joint surfaces, finished with corrosion-resistant nickel, suitable for use with spherical joints
- Spring wire design: four touch points for stability, secure swing clamp, stainless steel
Specifications
Specifications
| Style | Fork Design |
| Quantity | 5/Pk. |
| Type | Joint Clamp |
| Material | Nickel-plated Metal |
| For Use With (Equipment) | Glassware with flat flange and spherical joints |