Learn More
Quickfit™ Metal Clamps for Spherical Joints
Product Code. 12939383
Change view
Click to view available options
For Use With (Equipment):
Spherical joint size S13
Spherical joint size S19
Spherical joint size S35
Unit Size:
Pack of 2
Pack of 5
Product Code. For Use With (Equipment) unitSize
12939383 Spherical joint size S19 Pack of 5
12949383 Spherical joint size S35 Pack of 2
11322015 Spherical joint size S13 Pack of 5
3 options
Product Code. 12939383
Brand: Quickfit JC19
Finished with corrosion-resistant nickel for durability. Quickfit™ Metal Clamps for Spherical Joints are designed to secure corresponding Quickfit™ glassware with ball and socket style joints.
  • Suitable for high temperature work
  • Spring loaded to ensure balanced positive pressure to both the ball and socket joint surface
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5/Pk.
Type Spherical Joint Clamp
Material Nickel-plated Metal
For Use With (Equipment) Spherical joint size S19