Learn More
Quickfit™ Metal Clamps for Spherical Joints
Product Code. 11322015
Click to view available options
For Use With (Equipment):
Spherical joint size S13
Spherical joint size S19
Spherical joint size S35
Unit Size:
Pack of 2
Pack of 5
Product Code. 11322015
Brand: Quickfit JC13
Only null left
Add to basket
Add to basket
Finished with corrosion-resistant nickel for durability. Quickfit™ Metal Clamps for Spherical Joints are designed to secure corresponding Quickfit™ glassware with ball and socket style joints.
  • Suitable for high temperature work
  • Spring loaded to ensure balanced positive pressure to both the ball and socket joint surface
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5/Pk.
Type Spherical Joint Clamp
Material Nickel-plated Metal
For Use With (Equipment) Spherical joint size S13