Learn More
Quickfit Jointed Ware™ PTFE Washers
For use with Quickfit™ glass screwthreads
Brand: Quickfit QW28/18
332.50 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.01100kg
Description
- For use with Quickfit™ glass screwthreads
- Melamine screwcaps with aperture at the top
- Silicone rubber rings with PTFE washers
- To determine what components are required: i) Check the size of the glass screw thread (size designation refers to O. D. of thread inmm) ii) Check the O. D. of the sampling tube, thermometer, probe etc. , that is to be passed through the screw cap iii) Consult the table below remembering to order relevant cap, washer and ring separately
Specifications
For Use with Quickfit Glass Screw threads | |
Washer | |
10 Pack | |
PTFE |