Learn More
Quickfit™ Hexagonal Hollow-blown Glass Stopper
Click to view available options
Standard Taper Joint:
10/19
14/23
45/40
NS 12/21
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 7/16
Unit Size:
Pack of 10
Pack of 5
Description
Ideally suited for use with volumetric flasks.
- Lower closed end of stopper features drip-tip to ensure adhered liquid is returned to flask
- Hexagonal head prevents stopper from rolling and provides good grip
- Flat head allows stopper to be stood on end, thus preventing contamination of ground surface
- Hollow-blown to reduce weight
Specifications
Specifications
| For Use With (Equipment) | Volumetric flasks |
| Quantity | 5/Pk. |
| Standard Taper Joint | 45/40 |
| Material | Borosilicate Glass |