Learn More
Quickfit™ Hexagonal Hollow-blown Glass Stopper
Ideally suited for use with volumetric flasks.
Brand:  Quickfit TRF690
622.25 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.06000kg
 View more versions of this product
Product Code. 11339093
  • 655.00 NOK / Pack of 10
Estimated Shipment: 19-06-2024
Description
Ideally suited for use with volumetric flasks.
  • Lower closed end of stopper features 'drip-tip' to ensure adhered liquid is returned to flask
  • Hexagonal head prevents stopper from rolling and provides good grip
  • Flat head allows stopper to be stood on end, thus preventing contamination of ground surface
  • Hollow-blown to reduce weight
Specifications
Specifications
Hollow-Blown Glass
10/Pk.
Volumetric flasks
7/16
Glass
NS 7/16