Learn More
Quickfit™ Glass Büchner Funnel with Glass Sintered Disc
Product Code. 12008483
Change view
Click to view available options
Capacity (Metric):
125 mL
500 mL
Diameter (Metric) Disc:
55mm
95mm
Cone Size:
19/26
24/29
29/32
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Disc Cone Size unitSize
12008483 500 mL 95mm 24/29 Each
12425386 125 mL 55mm 19/26 Each
12098473 125 mL 55mm 24/29 Each
15567960 125 mL 55mm 29/32 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12008483 Supplier Quickfit Supplier No. SF3B33
Only null left
Add to basket
Add to basket
Used in place of conventional filter papers that would be destroyed by acids and alkalis. Quickfit™ Glass Büchner Funnel with Sintered Glass Disc is manufactured from chemical-resistant borosilicate glass. Integral ground glass cone on stem connects to Quickfit™ Büchner flasks and vessels.
  • Büchner funnel with glass sintered disc
  • Borosilicate glass construction for high resistance to chemical attack
  • Useful where conventional filter papers would be destroyed by acids and alkalis
  • Integral ground glass cone on stem for connecting to Quickfit™ Büchner flasks etc
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass
Cone Size 24/29
For Use With (Application) Bases or acids that can destroy conventional filter papers
Diameter (Metric) Disc 95mm
Includes Sintered glass disc, ground glass cone
Porosity 3
Pore Size 16 to 40μm
Product Type Buchner Funnel