Learn More
Pyrex™ Quickfit™ Glass Key and Stem with Cone
Product Code. 11361875
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Cone Size:
14/23
19/26
Stopcock:
2.0mm bore
2.5mm bore
4.0mm bore
Unit Size:
Each
Pack of 5
Product Code. Capacity (Metric) Cone Size Stopcock unitSize
11361875 250 mL 19/26 2.5mm bore Each
11341875 100 mL 14/23 2.5mm bore Each
11351875 100 mL 19/26 2.5mm bore Each
11331875 50 mL 14/23 2.0mm bore Each
15517790 500 mL 19/26 4.0mm bore Pack of 5
15537790 1000 mL 19/26 4.0mm bore Pack of 5
6 options
Product Code. 11361875
Brand: Pyrex™ D1/32
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Add liquid to most Quickfit™ flasks and assemblies. Pyrex™ Quickfit™ Glass Key and Stem with Cone includes stopcock with interchangeable glass key.
  • Complies with ISO 4800
  • Funnel stem with ground glass cone for easy incorporation into Quickfit™ flasks and assemblies
  • Stopcock has interchangeable glass key
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Material Pyrex Glass, Borosilicate Glass
Certifications/Compliance ISO 4800
Cone Size 19/26
Includes Ground glass cone, stopcock with glass key
Stopcock 2.5mm bore
Product Type Dropping funnel