Learn More
Quickfit™ Expansion Socket Adapter
Código de producto. 11389093
Change view
Click to view available options
Cone Size:
10/19
14/23
19/26
24/29
29/32
Standard Taper Joint No.:
14/23
19/26
24/29
29/32
34/35
40/38
Tamaño de la unidad:
Each
Pack of 2
Código de producto. Cone Size Standard Taper Joint No. unitSize
11389093 29/32 40/38 Each
12146220 14/23 19/26 Pack of 2
10427820 19/26 34/35 Pack of 2
12495436 14/23 24/29 Pack of 2
12156220 19/26 24/29 Pack of 2
11928044 14/23 29/32 Pack of 2
12614901 10/19 14/23 Pack of 2
11996124 19/26 29/32 Pack of 2
10881391 24/29 29/32 Pack of 2
12425446 29/32 34/35 Pack of 2
10146454 24/29 34/35 Pack of 2
11 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11389093
Marca: Quickfit XA64
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Este artículo no se puede devolver. Vea la política de devoluciones
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
  • Useful for the connection of dissimilar sized ground glass joints
  • Large sized socket at the top and small sized cone at the base
TRUSTED_SUSTAINABILITY
Specifica
Cone Size 29/32
For Use With (Application) Useful for the connection of dissimilar sized ground Glass joints
Material Borosilicate Glass
Product Type Expansion adapter
Standard Taper Joint No. 40/38
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.