Learn More
Quickfit™ Chromatography Column with Rotaflo™ Stopcock
Product Code. 11318074
Change view
Click to view available options
Length (Metric):
200 mm
300 mm
400 mm
500 mm
Diameter (Metric) Inner:
10 mm
20 mm
26 mm
40 mm
Connector Types:
14/23 Socket
19/26 Socket
24/29 Socket
40/38 Socket
Includes:
GP Rotaflo 3mm Bore Stopcock
GP Rotaflo 6mm Bore Stopcock
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Connector Types Includes unitSize
11318074 300 mm 20 mm 19/26 Socket GP Rotaflo 3mm Bore Stopcock Each
12011292 500 mm 26 mm 24/29 Socket GP Rotaflo 3mm Bore Stopcock Each
12478826 400 mm 10 mm 14/23 Socket GP Rotaflo 3mm Bore Stopcock Each
12468836 500 mm 40 mm 40/38 Socket GP Rotaflo 6mm Bore Stopcock Each
12081262 200 mm 10 mm 14/23 Socket GP Rotaflo 3mm Bore Stopcock Each
12498826 500 mm 20 mm 19/26 Socket GP Rotaflo 3mm Bore Stopcock Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11318074 Supplier Quickfit Supplier No. CR20/30
From Chemicals to Chromatography
Use promo code 27991
View all promotions
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Manufactured from Pyrex™ borosilicate glass. Quickfit™ Chromatography Column with Rotaflo™ Stopcock includes a 0 porosity chemically resistant glass sintered disc. PTFE Stopcock provides fine control of eluent flow. Ground glass socket accepts Quickfit™ reservoirs.
  • With PTFE Rotaflo™ Stopcock for fine control of eluent flow
  • Chemically resistant glass sintered disc - porosity 0
  • Ground glass socket accepts Quickfit™ reservoirs
  • Manufactured from Pyrex™ borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Pack
Diameter (Metric) Inner 20 mm
Type Chromatography Column
Length (Metric) 300 mm
Material Pyrex™ Borosilicate Glass
Connector Types 19/26 Socket
Includes GP Rotaflo 3mm Bore Stopcock