Learn More
Quickfit™ Borosilicate Glass Vigreux Fractionating Column
Click to view available options
Height (Metric):
360 mm
600 mm
Unit Size:
Pack of 5
Description
- Popular, easy to use column
- Vigreux type with combination of horizontal and angled indentations
- Horizontal indentations ensure intimate vapour and liquid contact
- Angled indentations redistribute liquid from walls to center of column
Specifications
Specifications
| Includes | 2 NS 29/32 standard ground joint |
| Height (Metric) | 360 mm |
| Length (Metric) | 360mm |
| Type | Vigreux Column |
| Material | Glass |
| Quantity | 1Each |
| Standard Taper Joint | NS 29/32 |