Learn More
Quickfit™ Three-Vertical-Neck Round-Bottom Flask
Product Code. 15507890
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
3000 mL
4 L
5 L
6 L
Socket Joint:
24/29 (Centre), 19/26 (side)
29/32 (Center), 14/23 (Sides)
29/32 (Center), 29/32 (Sides)
29/32 (Centre), 19/26 (side)
34/35 (Centre), 19/26 (side)
34/35 (Centre), 24/29 (side)
45/40 (Center), 29/32 (sides)
45/40 (Centre), 29/32 (side)
Centre: 29/32, Side: 19/26
Unit Size:
Each
Pack of 5
Product Code. 15507890
Brand: Quickfit FR3L/5S/22P
Only null left
Add to basket
Add to basket
Includes three necks for easy addition of measurement tools. The Quickfit™ Three-Vertical-Neck Round-Bottome Flask is made from borosilicate glass.
  • Fitted with three Quickfit™ precision-ground sockets - One vertical centre neck - Two angled side necks
  • Quickfit™ ground glass sockets are manufactured to an accuracy which surpasses the requirements of ISO 383
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 3000 mL
Material Borosilicate Glass
Height (Metric) 249 mm
No. per Case 3
Socket Joint 34/35 (Centre), 19/26 (side)
Bottom Shape Round