Learn More
Quickfit™ Two-Neck Pear-Shaped Flask
Click to view available options
:
Capacity: 50mL; Socket: 14/23 (Centre), 14/23 (side)
Description
Side neck useful for introducing gas inlet tubes and thermometers. The Quickfit Two-Neck Pear-Shaped Flask is made from borosilicate glass.
- With highest-quality ground glass socket
- Side neck useful for introducing gas inlet tubes and thermometers etc., into the flask
- Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack
- With one vertical center neck and one angled side neck
Specifications
Specifications
Capacity (Metric) | 50 mL |
Material | Borosilicate Glass |
Height (Metric) | 115 mm |
Neck Style | One vertical centre neck and one angled side neck |
Socket Joint | 14/23 (Centre), 14/23 (side) |
Bottom Shape | Pear |
Documents
Product Information
Product Information
Product information
File size: 2.8 MB
Product Content Correction
Product Title
Quickfit™ Two-Neck Pear-Shaped Flask >