Learn More
QuickFit™ Borosilicate Glass Test Tubes with Ground Socket
Product Code. 11382045
Change view
Click to view available options
Diameter (Metric):
14 mm
17 mm
19 mm
23 mm
29 mm
41 mm
Capacity (Metric):
5 mL
10 mL
11 mL
15 mL
19 mL
31 mL
54 mL
75 mL
195 mL
Unit Size:
Each
Pack of 5
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Capacity (Metric) unitSize
11382045 19 mm 11 mL Pack of 5
11352055 29 mm 75 mL Pack of 5
10747645 19 mm 19 mL Pack of 5
11302055 19 mm 19 mL Each
10715101 41 mm 195 mL Pack of 5
12475316 14 mm 5 mL Pack of 5
11392045 19 mm 15 mL Pack of 5
11342055 29 mm 54 mL Pack of 5
10074474 23 mm 19 mL Pack of 5
10497400 23 mm 31 mL Pack of 5
12569795 17 mm 10 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 11382045 Supplier Quickfit Supplier No. MF24/1
Manufactured from borosilicate glass for excellent chemical and thermal shock resistance. Quickfit™ Borosilicate Glass Test Tubes with Ground Socket are ideal for mixing and heating chemicals in a lab.
  • Presicion-ground socket ensures exact fit for stopper
  • Test tube sizes correspond with Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers
  • Order tubes and stoppers separately
TRUSTED_SUSTAINABILITY
Specifications
Design Type Ground Socket
Description Stopper Size: 14/23
Capacity (Metric) 11 mL
Length (Metric) 100 mm
Material Borosilicate Glass
For Use With Quickfit Hollow Stopper, Fisher Scientific product code 10155824
Quantity 5 Pack
Diameter (Metric) 19 mm