Learn More
QuickFit™ Borosilicate Glass Test Tubes with Ground Socket
Product Code. 10074474
Click to view available options
Diameter (Metric):
14 mm
17 mm
19 mm
23 mm
29 mm
41 mm
Capacity (Metric):
5 mL
10 mL
11 mL
15 mL
19 mL
31 mL
54 mL
75 mL
195 mL
Unit Size:
Each
Pack of 5
Product Code. 10074474
Brand: Quickfit MF24/2
Only null left
Add to basket
Add to basket
Manufactured from borosilicate glass for excellent chemical and thermal shock resistance. Quickfit™ Borosilicate Glass Test Tubes with Ground Socket are ideal for mixing and heating chemicals in a lab.
  • Presicion-ground socket ensures exact fit for stopper
  • Test tube sizes correspond with Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers
  • Order tubes and stoppers separately
TRUSTED_SUSTAINABILITY
Specifications
Design Type Ground Socket
Description Stopper Size: 19/26
Capacity (Metric) 19 mL
Length (Metric) 100 mm
Material Borosilicate Glass
For Use With Quickfit Hollow Stopper, Fisher Scientific product code 12568855
Quantity 5 Pack
Diameter (Metric) 23 mm