Learn More
- Home
- Products
- Flasks
- Boiling Flasks
- Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Product Code. 15537840| Product Code. | Capacity (Metric) | Height (Metric) | unitSize | Socket Joint |
|---|---|---|---|---|
| 15537840 | 4 L | 260 mm | Pack of 5 | - |
| 12498936 | 1000 mL | 170 mm | Pack of 2 | 24/29 |
| 12498926 | 100 mL | 95 mm | Pack of 5 | 19/26 |
| 12418936 | 100 mL | 103 mm | Pack of 5 | 24/29 |
| 12428936 | 100 mL | 103 mm | Pack of 5 | 29/32 |
| 10243930 | 150 mL | 110 mm | Pack of 5 | 24/29 |
| 10697445 | 150 mL | 115 mm | Pack of 5 | 34/35 |
| 12428946 | 250 mL | 130 mm | Pack of 5 | 19/26 |
| 10309754 | 250 mL | 130 mm | Pack of 5 | 29/32 |
| 10831041 | 250 mL | 130 mm | Pack of 5 | 34/35 |
| 12509885 | 500 mL | 140 mm | Pack of 5 | 24/29 |
| 12418956 | 500 mL | 145 mm | Pack of 5 | 29/32 |
| 10851201 | 50 mL | 85 mm | Pack of 5 | 19/26 |
| 12661992 | 50 mL | 85 mm | Pack of 5 | 24/29 |
| 11980885 | 250 mL | 130 mm | Pack of 5 | 24/29 |
| 15527840 | 2000 mL | 215 mm | Pack of 5 | 34/35 |
| 12408946 | 1000 mL | 187 mm | Pack of 2 | 29/32 |
| 15567830 | 150 mL | 105 mm | Pack of 20 | 19/26 |
| 15577830 | 150 mL | 110 mm | Pack of 20 | 29/32 |
| 15517840 | 2000 mL | 230 mm | Pack of 5 | 29/32 |
| 15547840 | 500 mL | 135 mm | Pack of 20 | 19/26 |
| 15567840 | 500 mL | 155 mm | Pack of 20 | 34/35 |
| 15557830 | 100 mL | 103 mm | Pack of 10 | - |
| 15597830 | 1000 mL | 175 mm | Pack of 10 | 34/35 |
| 15507840 | 2000 mL | 205 mm | Pack of 5 | 24/29 |
Product Code. 15537840
Brand: Quickfit FF4L/7S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
- Complies with ISO 1773 and ISO 4797
- With highest quality Quickfit™ ground glass socket
- Borosilicate glass for highest resistance to thermal shock and chemical corrosion
- Flat base allows use with hotplates
Specifications
Specifications
| Certifications/Compliance | ISO 1773, ISO 4797 |
| Capacity (Metric) | 4 L |
| Material | Borosilicate Glass |
| Height (Metric) | 260 mm |
| Neck Style | Short |
| Bottom Shape | Flat |