Learn More
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Product Code. 12418936
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
150 mL
250 mL
500 mL
1000 mL
2000 mL
4 L
Height (Metric):
85 mm
95 mm
103 mm
105 mm
110 mm
115 mm
130 mm
135 mm
140 mm
145 mm
155 mm
170 mm
175 mm
187 mm
205 mm
215 mm
230 mm
260 mm
Socket Joint:
19/26
24/29
29/32
34/35
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Pack of 5
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) Socket Joint unitSize
12418936 100 mL 103 mm 24/29 Pack of 5
12498936 1000 mL 170 mm 24/29 Pack of 2
12498926 100 mL 95 mm 19/26 Pack of 5
12428936 100 mL 103 mm 29/32 Pack of 5
10243930 150 mL 110 mm 24/29 Pack of 5
10697445 150 mL 115 mm 34/35 Pack of 5
12428946 250 mL 130 mm 19/26 Pack of 5
10309754 250 mL 130 mm 29/32 Pack of 5
10831041 250 mL 130 mm 34/35 Pack of 5
12509885 500 mL 140 mm 24/29 Pack of 5
12418956 500 mL 145 mm 29/32 Pack of 5
10851201 50 mL 85 mm 19/26 Pack of 5
12661992 50 mL 85 mm 24/29 Pack of 5
11980885 250 mL 130 mm 24/29 Pack of 5
15527840 2000 mL 215 mm 34/35 Pack of 5
12408946 1000 mL 187 mm 29/32 Pack of 2
15567830 150 mL 105 mm 19/26 Pack of 20
15577830 150 mL 110 mm 29/32 Pack of 20
15517840 2000 mL 230 mm 29/32 Pack of 5
15547840 500 mL 135 mm 19/26 Pack of 20
15567840 500 mL 155 mm 34/35 Pack of 20
15557830 100 mL 103 mm - Pack of 10
15597830 1000 mL 175 mm 34/35 Pack of 10
15507840 2000 mL 205 mm 24/29 Pack of 5
15537840 4 L 260 mm - Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
Product Code. 12418936 Supplier Quickfit Supplier No. FF100/3S
Only null left
Add to basket
Add to basket
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
  • Complies with ISO 1773 and ISO 4797
  • With highest quality Quickfit™ ground glass socket
  • Borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • Flat base allows use with hotplates
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 1773, ISO 4797
Capacity (Metric) 100 mL
Material Borosilicate Glass
Height (Metric) 103 mm
Neck Style Short
Socket Joint 24/29
Bottom Shape Flat