Learn More
Quickfit™ Medium Neck Flat Bottom Flask
Product Code. p-8002645
Change view
Click to view available options
Capacity (Metric):
100 mL
150 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 10
Pack of 2
Product Code. Capacity (Metric) unitSize
12458936 150 mL Pack of 2
10871031 250 mL Pack of 2
15547830 100 mL Pack of 10
15587830 1000 mL Pack of 10
15557840 500 mL Pack of 10
5 options
Product Code. 12458936
Brand: Quickfit FF150/3M
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Useful in reducing problems associated with splashing and frothing.
  • Made of Borosilicate glass
  • With longer neck than standard Quickfit flasks
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 1773, ISO 4797
Capacity (Metric) 150 mL
Material Borosilicate Glass
Height (Metric) 140 mm
Neck Style Medium