Learn More
Quickfit™ Borosilicate Glass Liebig Condenser
Description
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
- Complies with DIN 12576
- Popular water-jacketed condenser suitable for distillation and reflux applications
- With detachable plastic screwthread connectors for easy and safe attachment of rubber hose
Specifications
Specifications
| Certifications/Compliance | DIN 12576 |
| Description | Surface Area X 102m2: 2.00 |
| For Use With (Application) | Suitable for distillation and reflux applications |
| Length (Metric) Jacket | 400 mm |
| Length (Metric) | 530 mm |
| Standard Taper Joint No | 19/26 |
| Material | Borosilicate Glass |
| Type | Liebig Condenser |