Learn More
- Home
- Products
- Flasks
- Erlenmeyer Flasks
- Quickfit™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Quickfit™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Product Code. 10840861
Change view
Click to view available options
Capacity (Metric):
5 mL
25 mL
50 mL
100 mL
150 mL
250 mL
500 mL
1000 mL
2000 mL
Height (Metric):
54 mm
70 mm
85 mm
90 mm
100 mm
116 mm
118 mm
130 mm
175 mm
220 mm
270 mm
Diameter (Metric) Outer:
25 mm
42 mm
51 mm
64 mm
74 mm
85 mm
101 mm
130 mm
160 mm
Socket Joint:
14/23
19/26
24/29
29/32
34/35
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Pack of 5
26 product options available for selection
| Product Code. | Capacity (Metric) | Height (Metric) | Diameter (Metric) Outer | Socket Joint | unitSize |
|---|---|---|---|---|---|
| 10840861 | 250 mL | 130 mm | 85 mm | 24/29 | Pack of 5 |
| 12599205 | 150 mL | 116 mm | 74 mm | 29/32 | Pack of 5 |
| 12692052 | 50 mL | 85 mm | 51 mm | 19/26 | Pack of 5 |
| 10600701 | 25 mL | 70 mm | 42 mm | 14/23 | Pack of 5 |
| 10253540 | 150 mL | 118 mm | 74 mm | 19/26 | Pack of 5 |
| 10345340 | 50 mL | 90 mm | 51 mm | 24/29 | Pack of 5 |
| 10243880 | 100 mL | 100 mm | 64 mm | 19/26 | Pack of 5 |
| 12081682 | 250 mL | 130 mm | 85 mm | 29/32 | Pack of 5 |
| 10641861 | 25 mL | 70 mm | 42 mm | 19/26 | Pack of 5 |
| 10508980 | 100 mL | 100 mm | 64 mm | 24/29 | Pack of 5 |
| 10243400 | 1000 mL | 220 mm | 130 mm | 24/29 | Pack of 2 |
| 10743961 | 150 mL | 118 mm | 74 mm | 24/29 | Pack of 5 |
| 10315060 | 100 mL | 100 mm | 64 mm | 14/23 | Pack of 5 |
| 10263880 | 500 mL | 175 mm | 101 mm | 29/32 | Pack of 5 |
| 10651861 | 500 mL | 175 mm | 101 mm | 24/29 | Pack of 5 |
| 10851021 | 250 mL | 130 mm | 85 mm | 34/35 | Pack of 5 |
| 10496840 | 50 mL | 85 mm | 51 mm | 14/23 | Pack of 5 |
| 12569205 | 100 mL | 100 mm | 64 mm | 29/32 | Pack of 5 |
| 12478916 | 250 mL | 130 mm | 85 mm | 19/26 | Pack of 5 |
| 15577820 | 1000 mL | 220 mm | 130 mm | 29/32 | Pack of 10 |
| 15587820 | 1000 mL | 220 mm | 130 mm | - | Pack of 10 |
| 15597820 | 2000 mL | 270 mm | 160 mm | 29/32 | Pack of 5 |
| 15507830 | 2000 mL | 270 mm | 160 mm | 34/35 | Pack of 5 |
| 15517830 | 5 mL | 54 mm | 25 mm | - | Pack of 20 |
| 15527830 | 500 mL | 175 mm | 101 mm | 19/26 | Pack of 20 |
| 15537830 | 500 mL | 175 mm | 101 mm | - | Pack of 20 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
This item is not returnable.
View return policy
Product Code.
10840861
Supplier
Quickfit
Supplier No.
FE250/3
Spotlight Offers
Use promo code 28036
Use promo code 28036
Estimated Shipment: 09-12-2026
Log in to see stock.
Log in to see stock.
Add to basket
1222.27 NOK valid until 2026-08-31
BEST PRICE on Promo! Use promo code "28036" to get your promotional price.
BEST PRICE on Promo! Use promo code "28036" to get your promotional price.
Add to basket
This item is not returnable.
View return policy
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Glass Erlenmeyer Flask with Quickfit Ground Glass Socket complies to ISO 4797.
- Complies to ISO 4797
- Conical flasks with highest quality Quickfit™ ground glass socket
- Borosilicate glass for corrosion free performance and high resistance to thermal shock
- Designed for the estimation of arsenic and antimony
- Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 538nm or 5380.
Specifications
Specifications
| Certifications/Compliance | ISO 4797 |
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 130 mm |
| Includes | Ground socket |
| Shape | Conical |
| Socket Joint | 24/29 |
| Diameter (Metric) Outer | 85 mm |