Learn More
Quickfit™ Borosilicate Glass Double Surface Davies Condenser
Product Code. 11391855
Click to view available options
Standard Taper Joint No:
14/23
19/26
24/29
29/32
34/35
Length (Metric) Jacket:
160 mm
205 mm
Length (Metric):
275 mm
295 mm
300 mm
340 mm
345 mm
350 mm
360 mm
365 mm
Description:
Surface area x 102m2: 1.7
Surface area x 102m2: 2.3
Surface area x 102m2: 3.0
Unit Size:
Each
Pack of 2
Product Code. 11391855
Brand: Quickfit C5/11/SC
Only null left
Add to basket
Add to basket
Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Double Surface Davies Condenser is highly efficient with a large cooling surface. The inner cooling surface prevents vibration with use of the baffle effect, while the outer cooling surface prevents creep with low boiling point liquids. Includes detachable plastic screw thread connectors for easy and safe tubing attachment.
Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Double Surface Davies Condenser is highly efficient with a large cooling surface. The inner cooling surface prevents vibration with use of the baffle effect, while the outer cooling surface prevents creep with low boiling point liquids. Includes detachable plastic screw thread connectors for easy and safe tubing attachment.
  • Highly efficient condenser with large cooling surface
  • Inner cooling surface gives 'baffle' effect
  • Outer cooling surface prevents 'creep' with low boiling point liquids
  • With detachable plastic screw thread connectors for easy and safe attachment of flexible tubing
TRUSTED_SUSTAINABILITY
Specifications
Description Surface area x 102m2: 1.7
Length (Metric) Jacket 160 mm
Length (Metric) 275 mm
Standard Taper Joint No 14/23
Material Borosilicate Glass
Type Davies Condenser