Learn More
Quickfit™ Borosilicate Glass Cylindrical Culture Vessel
Description
Manufactured from temperature resistant borosilicate glass. QuickfitTM Borosilicate Glass Cylindrical Culture Vessel is ideal for fermentation and culture work. Autoclavable with precision ground flat-flange joint. For use with QuickfitTM flat-flange lids and retaining clips.
- Ideal for fermentation and culture work
- Manufactured from temperature resistant borosilicate glass
- Autoclavable
- With precision ground flat-flange joint
- For use with Quickfit™ flat-flange lids and retaining clips
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 10,000 mL |
| Material | Borosilicate Glass |
| For Use With (Application) | Quickfit™ flat flange lids and retaining clips, ideal for fermentation and culture work |
| No. per Pack | 1 Each |
| Height (Metric) | 375 mm |
| Diameter (Metric) | 227 mm |