Learn More
Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone
Description
Circulates coolant through a coil with a very large cooling surface. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone is ideal for use in the upright reflux mode where counter-current cooling is possible. Supplied with detachable plastic connectors for the attachment of rubber hose.
- Coolant is circulated through a coil having a very large cooling surface
- Ideal for use in the upright reflux mode where counter-current cooling is possible
- Supplied with detachable plastic connectors for the attachment of rubber hose
Specifications
Specifications
| For Use With (Application) | Ideal for use in the upright reflux mode where counter-current cooling is possible |
| Length (Metric) Jacket | 348 mm (Overall) 155 mm (Effective) |
| Standard Taper Joint No | 50/42 |
| Material | Borosilicate Glass |
| Type | Condenser Coil |