Learn More
Quickfit™ Borosilicate Glass Büchner Vacuum Filtration Funnels with Glass Sintered Disc
Product Code. 15598000
Change view
Click to view available options
Capacity (Metric):
30 mL
80 mL
125 mL
500 mL
Porosity:
1
2
3
4
Diameter (Metric) Disc:
40 mm
40mm
60mm
95 mm
95mm
Disc: 30mm
Unit Size:
Pack of 5
Product Code. Capacity (Metric) Porosity Diameter (Metric) Disc unitSize
15598000 80 mL 4 40mm Pack of 5
15728127 30 mL 4 Disc: 30mm Pack of 5
15528010 125 mL 4 60mm Pack of 5
15568000 80 mL 1 40mm Pack of 5
15738127 500 mL 2 95mm Pack of 5
15748127 500 mL 4 95mm Pack of 5
15508010 125 mL 1 60mm Pack of 5
15588000 80 mL 3 40 mm Pack of 5
15538010 500 mL 3 95 mm Pack of 5
9 options
Product Code. 15598000
Brand: Quickfit VB40/08M
Only null left
Add to basket
Add to basket
Useful where conventional filter papers would be destroyed by acids and alkalis
  • Manufactured from borosilicate glass for highest resistance to chemical attack
  • With Quickfit ground glass cone to allow direct filtration into Quickfit Büchner flasks
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 80 mL
Material Borosilicate Glass
Height (Metric) 52 mm
Certifications/Compliance ISO 3585, DIN 12217, ASTM E-438
Cone Size 24/29
For Use With (Application) For filtering solutions that can be filtered by filter papers which are not resistant to certain acids and bases
Diameter (Metric) Disc 40mm
Includes Quickfit™ ground Glass cone
Porosity 4
Product Type Buchner Vacuum Filtration Funnel