Learn More
DWK Life Sciences™ Quick Release Clamp, from Stainless steel, with retaining clip
Shop All DWK Life Sciences Products
missing translation for 'orderingAttributeHoverText'
Size:
DN 100
DN 120
DN 150
DN 200
DN 60
missing translation for 'unitSize'
Each
Beschreibung
The quick release clamp closes flat flange vessels and secures the lid. Slipping or unchecked opening is not possible. The closure is made of high-quality Stainless steel and has three holding segments.
- With retaining clip
- Quick closing with quick-release fastener
Spezifikation
Spezifikation
| Quantity | 1 Ea. |
| Material | Stainless Steel |
| For Use With (Equipment) | Quick Release Clamp for Closure |
| For Use With (Application) | Quick release clamp for closure; Accessories for flat flange vessels |
| Size | DN 100 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur