Learn More
Quercetin 3-O-(6' '-O-Malonyl)-b-D-glucoside, TRC
Specifications
Specifications
| Chemical Name or Material | Quercetin 3-O-(6''-O-Malonyl)-b-D-glucoside |
| CAS | 96862-01-0 |
| Molecular Formula | C24 H22 O15 |
| Quantity | 1 mg |
| Synonym | 3-[[6-O-(2-Carboxyacetyl)-β-D-glucopyranosyl]oxy]-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, Quercetin 3-O-(6-O-malonyl)-β-D-glucoside |
| InChI Formula | InChI=1S/C24H22O15/c25-9-4-12(28)17-13(5-9)37-22(8-1-2-10(26)11(27)3-8)23(19(17)33)39-24-21(35)20(34)18(32)14(38-24)7-36-16(31)6-15(29)30/h1-5,14,18,20-21,24-28,32,34-35H,6-7H2,(H,29,30)/t14-,18-,20+,21-,24+/m1/s1 |
| SMILES | O[C@H]1[C@H](O)[C@@H](COC(=O)CC(=O)O)O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@@H]1O |
| IUPAC Name | 3-[[(2R,3S,4S,5R,6S)-6-[2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4-oxochromen-3-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3-oxopropanoic acid |
| Molecular Weight (g/mol) | 550.423 |
| Formula Weight | 550.096 |