Learn More
Invitrogen™ Qubit™ microRNA Assay Kits
Description
Achieve accurate and precise quantification of small RNA (∼20 nucleotides or base pairs) with Qubit miRNA (microRNA) Assay Kits. These miRNA quantification kits enable quick detection of all types of small RNA, including microRNA and siRNA, and both single stranded and double stranded RNA. The assay is highly selective for small RNA over rRNA or large mRNA, and can tolerate contaminants such as salts, solvents, or detergents.
The Qubit microRNA Assay Kit provides concentrated assay reagent, dilution buffer, and pre-diluted miRNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using the Qubit Fluorometer.
Notes
• Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer
Specifications
Specifications
| Detection Method | Fluorescence |
| For Use With (Equipment) | Qubit Fluorometer |
| For Use With (Application) | microRNA Quantitation |
| No. of Reactions | 500 Reactions |
| Product Line | Qubit |
| Product Type | microRNA Assay Kit |
| Quantity | 500 assays |
| Shipping Condition | Room Temperature |
| Excitation/Emission | 498/518 |