Learn More
Invitrogen™ Qubit™ microRNA Assay Kits
Product Code. 15241038
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
1 set
100 assays
Product Code. No. of Reactions unitSize
15241038 100 Reactions 100 assays
15401563 500 Reactions 1 set
2 options
Product Code. 15241038
Brand: Invitrogen™ Q32880
Edge
Achieve accurate miRNA quantitation, even in the presence of rRNA, large mRNA, or contaminants, with the Qubit microRNA Assay Kit, which offers 100 and 500 assay formats.
Achieve accurate and precise quantification of small RNA (∼20 nucleotides or base pairs) with Qubit miRNA (microRNA) Assay Kits. These miRNA quantification kits enable quick detection of all types of small RNA, including microRNA and siRNA, and both single stranded and double stranded RNA. The assay is highly selective for small RNA over rRNA or large mRNA, and can tolerate contaminants such as salts, solvents, or detergents.
The Qubit miRNA Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of microRNA samples. This assay is highly selective for miRNA over rRNA or large mRNA (>1000 bp). Depending on sample volume, the assay kit is designed to be accurate for initial miRNA sample concentrations of 0.025 to 150 ng/μL, providing a detection range of 0.5−150 ng.
The Qubit microRNA Assay Kit provides concentrated assay reagent, dilution buffer, and pre-diluted miRNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using the Qubit Fluorometer.
Notes
• Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer
TRUSTED_SUSTAINABILITY
Specifications
Detection Method Fluorescence
For Use With (Equipment) Qubit Fluorometer
For Use With (Application) microRNA Quantitation
No. of Reactions 100 Reactions
Product Line Qubit
Product Type microRNA Assay Kit
Quantity 100 assays
Shipping Condition Room Temperature
Excitation/Emission 498/518