Learn More
Invitrogen™ Qubit™ Fluorometer USB Flash Drive
Product Code. 15843764
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15843764 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15843764 Supplier Invitrogen™ Supplier No. Q46009
Estimated Shipment: 14-12-2026
The Qubit Fluorometer USB Flash Drive is designed for the transfer of data from the Qubit Fluorometer.
The Qubit Fluorometer USB Flash Drive is designed for the transfer of data from the Qubit Fluorometer. Only the Qubit 2.0, 3, and 4 fluorometers have an available USB port. Data exported from the Qubit Fluorometer are stored in CSV format and can be opened by programs such as Microsoft Excel.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Qubit
Shipping Condition Room Temperature
Type USB Flash Drive
Quantity 1 each
For Use With (Equipment) Qubit Fluorometer