Learn More
Invitrogen™ Qubit™ Fluorometer International Power Supply
Product Code. 15940493
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15940493 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15940493 Supplier Invitrogen™ Supplier No. A36204
 Restricted Item
Estimated Shipment: 10-12-2026
The Qubit Fluorometer International Power Supply is designed for use with either the Qubit 3 or Qubit 4 Fluorometer.
The Qubit Fluorometer International Power Supply is designed for use with either the Qubit 3 or Qubit 4 Fluorometer. This power supply should not be used with the original or Qubit 2.0 Fluorometer. The product includes a power cord and four interchangeable adaptors and enables the Qubit to be plugged into outlets around the world.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Qubit
Shipping Condition Room Temperature
Type Power Supply
Quantity 1 each
For Use With (Equipment) Qubit 3 Fluorometer, Qubit 4 Fluorometer