Learn More
Macherey-Nagel™ Quat Test Strip
Product Code. 10398404 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
100 Pack
Unit Size:
Pack of 100
Product Code. Quantity unitSize
10398404 100 Pack Pack of 100
1 options
Product Code. 10398404
Brand: Macherey-Nagel™ 91337
Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
  • Rapid: most tests can be carried out in 10s to 120s
  • Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
  • Precise: color charts adjusted and checked using certified standard solutions
  • Shelf life: 2. 5 years, store below 30°C
Order Info
Graduations
Specifications
Format Semi Quantitative Test Strips
For Use With (Application) For the rapid and reliable determination of quaternary ammonium compounds (QUAT) in solutions
Graduations 0 to 1000mg/L
Quantity 100 Pack
Type Quat Test Strip