Learn More
Motic™ Quartz Wedge For Motic™ TEC POL Microscopes
Description
For use with the following Motic™ Microscopes:
- Panthera TEC POL Binocular
- Panthera TEC POL Trinocular
- Panthera TEC POL Epi Binocular
- Panthera TEC POL Epi Trinocular
- Panthera TEC POL Digital
- Panthera TEC POL Epi Digital
- Panthera TEC POL (i)
Specifications
Specifications
| Type | Quartz Wedge |
| Color | Black |
| For Use With (Equipment) | Motic TEC POL Microscopes |