Learn More
Hellma™ Quartz Suprasil Ultramicro Cell
Product Code. 12594557
Change view
Click to view available options
Capacity (Metric):
5 μL
45 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12594557 5 μL Each
11756204 45 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12594557 Supplier Hellma™ Supplier No. 105210101540
  • ONLINE EXCLUSIVE PRICE
    8970.00 NOK
    8730.40 NOK / Pack of 1
    Save 239.60 NOK
    2% Off
Estimated Shipment: 05-02-2027
Specifications
Type Ultra-micro Cell
Capacity (Metric) 5 μL
Material Quartz Suprasil
Height (Metric) 45 mm
Width (Metric) 12.5 mm