Learn More
Hellma™ Suprasil™ Quartz Semi-Micro Cell Cuvettes
Product Code. 10518881
Change view
Click to view available options
Height (Metric):
45 mm
46 mm
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
10518881 46 mm Each
11786284 45 mm Pack of 2
10739491 45 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10518881 Supplier Hellma™ Supplier No. 114F1040
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Suprasil™ Quartz Semi-Micro Cell Cuvettes are manufactured from quartz, which is chemically very resistant and maintains its shape, even at high temperatures up to approximately 1000°C.
  • High UV transmission
  • Low thermal expansion
Specifications
Type Semi-micro Cell
Capacity (Metric) 1.4 mL
Material Quartz Suprasil
Closure Material PTFE
Width (Metric) Interior 4mm
Quantity 1Pack
Path Length 10 mm
Wavelength Range 260 to 2500 nm
Height (Metric) 46 mm
Width (Metric) 12.5 mm
For Use With (Application) Fluorescence Measurement