Learn More
Hellma™ Quartz Suprasil Micro Cells
Product Code. 10363981
Click to view available options
Path Length:
5 mm
50 mm
10 x 10 mm
Unit Size:
Each
Product Code. 10363981
Brand: Hellma™ 1105040
Specifications
Type Micro Cell
Capacity (Metric) 17.5 mL
Material Quartz Suprasil
Closure Material PTFE
Width (Metric) Interior 9.5mm
Quantity 1Each
Path Length 50 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 46 mm
Width (Metric) 12.5 mm