Learn More
Hellma™ Quartz Suprasil Micro Cells
Product Code. 10209951
Change view
Click to view available options
Path Length:
5 mm
50 mm
10 x 10 mm
Unit Size:
Each
Product Code. Path Length unitSize
10209951 5 mm Each
10363981 50 mm Each
10787181 10 x 10 mm Each
3 options
Product Code. 10209951
Brand: Hellma™ 110540
Only null left
Specifications
Type Micro Cell
Capacity (Metric) 1.75 mL
Material Quartz Suprasil
Closure Material PTFE
Width (Metric) Interior 9.5mm
Quantity 1Each
Path Length 5 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 46 mm
Width (Metric) 12.5 mm