Learn More
Hellma™ Quartz Suprasil Macro Absorption Cells
Click to view available options
Capacity (Metric):
700 μL
Unit Size:
Each
Specifications
Specifications
| Type | Macro Absorption Cell |
| Capacity (Metric) | 700 μL |
| Material | Quartz Suprasil |
| Width (Metric) Interior | 9.5mm |
| Quantity | 1 Each |
| Path Length | 2 mm |
| Height (Metric) | 52 mm |
| Width (Metric) | 12.5 mm |