Learn More
Hellma™ Quartz Suprasil™ Micro Absorption Cells
Product Code. 10281851
Change view
Click to view available options
Capacity (Metric):
0.4 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10281851 0.4 mL Each
1 options
Product Code. 10281851
Brand: Hellma™ 1151040
Only null left
Add to basket
Add to basket
Thanks to their stability, maximum precision and reliability when used for absorbance measurements, Hellma cuvettes work exceptionally well in a wide range of areas in the lab.
  • Quartz glass high performance of the highest purity and homogeneity is used for the window
  • Guaranteed transmission of at least 85% from 200nm to 3500nm depending on material
  • Extremely high parallelism of the windows with a maximum tolerance of ±0.01mm
  • Exceptional optical path length accuracy down to 0.003mm for high dimensional accuracy and reproducible measurement results
  • Unique surface flatness of the optical windows of 0.001mm
  • Very high temperature stability and chemical resistance due to thermal bonding of individual components
Specifications
Type Absorption Cell
Description Clear side walls and base
Capacity (Metric) 0.4 mL
Material Quartz Suprasil
Width (Metric) Interior 2mm
Path Length 10 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 40 mm
Width (Metric) 12.5 mm