Learn More
Thermo Scientific Chemicals Quartz disc, fused, 50.8mm (2.0in) dia x 3.18mm (0.13in) thick
Product Code. 15436455
Click to view available options
Diameter (Metric):
50.8 mm
76.2 mm
Thickness:
1.59 mm
3.18 mm
Unit Size:
Each
Product Code. 15436455
Brand: Thermo Scientific Chemicals 042298.KS
Request Bulk or Custom Format
Manufactured by fusing naturally occurring crystalline silica
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mineral, Quartz Disc
Description Form: Ground and polished
Diameter (Metric) 50.8 mm
Thickness 1.59 mm