Learn More
Thermo Scientific Chemicals Quartz, cover slip for microscope slide, fused, 25.4x25.4x0.15-0.25mm (1.0x1.0x0.006-0.01in)
Product Code. 15403895
Change view
Click to view available options
Quantity:
1/Ea.
5/Ea.
Dimensions (L x W):
25.4 mm
25.4 x 25.4 mm
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Dimensions (L x W) Unit Size
15403895 5/Ea. 25.4 mm Pack of 5
15473885 1/Ea. 25.4 x 25.4 mm Each
15483885 5/Ea. 25.4 x 25.4 mm Pack of 5
15493885 1/Ea. 25.4 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15403895 Supplier Thermo Scientific Chemicals Supplier No. 043211.KJ
  • ONLINE EXCLUSIVE PRICE
    3565.00 NOK
    2782.65 NOK / Pack of 5
    Save 782.35 NOK
    21% Off
In Stock
Add to basket
For use with microscope quartz slides made by fusing naturally occurring crystalline silica
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Cover slips for use with microscope quartz slides
TRUSTED_SUSTAINABILITY
Specifications
Thickness (Metric) 0.15 to 0.25 mm
Description Cover Slips for use with Microscope Quartz Slide
Diameter (Metric) 25.4mm
Quantity 5/Ea.
Dimensions (L x W) 25.4 mm