Learn More
Velp Scientifica™ Quartz Ash Insert
Product Code. 15389574 Shop All Velp Products
Change view
Click to view available options
Type:
Quartz Ash Insert
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15389574 Quartz Ash Insert Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15389574 Supplier Velp Scientifica™ Supplier No. A00000161
  • ONLINE EXCLUSIVE PRICE
    292.58 NOK
    257.37 NOK / Pack of 1
    Save 35.21 NOK
    12% Off
Estimated Shipment: 23-11-2026
Velp Scientifica™ Quartz Ash Insert for use with Dumas Nitrogen Analyzer NDA 701 and 702.
  • For use with NDA 701 and 702
Specifications
Type Quartz Ash Insert
For Use With Dumas Nitrogen Analyzer
Weight (Metric) 0.18 kg