Learn More
Applied Biosystems™ QuantStudio™ 3D Digital PCR Chip Adapter Kit for Flat Block Thermal Cycler
Shop All Applied Biosystems ProductsDescription
QuantStudio™ 3D Digital PCR Chips in the dual flat blocks of the GeneAmp™ 9700 or ProFlex™ thermal cyclers. The Adaptor Kit includes two block adaptors, as well as two compression pads that sit atop the chips during thermal cycling.
Specifications
Specifications
| Content And Storage | Contents: • 2 chip adapters • 2 thermal pads |
| Format | 3D Digital PCR Chips |
| PCR Method | Digital PCR |
| For Use With (Equipment) | GeneAmp 9700, GeneAmp™ PCR System 9700, Dual Flat Block, ProFlex™ 2x Flat PCR System, ProFlex™ PCR System |
| Product Line | QuantStudio |
| Product Type | 3d Digital PCR Chip Adapter Kit |
| Quantity | 1 kit |
| For Use With (Application) | Real Time PCR (qPCR) |