Learn More
Macherey-Nagel™ Quantofix™ Test Strips
Product Code. 11396594 Shop All Macherey-Nagel Products
Click to view available options
Description:
Glutaraldehyde
LubriCheck
Peracetic Acid 500
Silver
Total Iron 1000
Range:
0, 0.5, 1.0, 1.5, 2.0, 2.5 % Glutaraldehyde
0, 1, 2, 3, 5, 7, 10g/L Ag
0, 15, 50, 75, 130, 200mmol/L KOH
0, 5, 20, 50, 100, 250, 500, 1000mg/L Fe2+/3+
0, 50, 100, 200, 300, 400, 500mg/L Peracetic Acid
Color Code:
Bright Orange to Magenta
White to Dark Red
Yellow to Blue
Yellow to Brown
Yellow to Green
For Use With (Application):
Determination of Glutaraldehyde
Determination of Peracetic Acid
Determination of Potassium Hydroxide
Determination of Total Iron 1000
Determination of silver
Unit Size:
Each
Pack of 100
Product Code. 11396594
Brand: Macherey-Nagel™ 91343
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Perform fast, easy analyses on the spot. Macherey-Nagel™ Quantofix™ Test Strips are ready to use semi-quantitative tests that change color depending on the concentration of the analyte in the sample. Read results using the color scale or with the Quantofix™ Relax.
Quantofix™ test strips meet all requirements of a modern rapid test. Analytical professionals as well as occasional testers appreciate the test strips for the fast and easy analysis directly at the point of interest. Often, these tests are used to quickly check whether important parameters are in the desired range. They deliver an immediate result and thus enable a fast response.
  • Rapid
    • Just Dip & Read
    • Results within seconds
    • Always ready for use
  • Easy
    • No calibration
    • No maintenance
    • No accessories
  • Reliable
    • Desiccant in the stopper for optimal protection of the strips against humidity
    • Color chart confirmed with traceable standards
    • Automatic evaluation with Quantofix™ Relax for safe documentation
Specifications
Certifications/Compliance CE
Color Bright orange to Magenta
Color Code Bright Orange to Magenta
Content And Storage 4°C to 30°C
Description Glutaraldehyde
Detectable Analytes Glutaraldehyde
Format Strip
Type Semi-qualitative Test Strips
For Use With (Application) Determination of Glutaraldehyde
No. of Reactions 100
Quantity 100 Test Strips
Range 0, 0.5, 1.0, 1.5, 2.0, 2.5 % Glutaraldehyde
Test Time 20 sec.