Learn More
Macherey-Nagel™ Quantofix™ Peroxide 100 Test Strip Kits
Product Code. 10159960 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color Indicator:
White to Blue
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Indicator Unit Size
10159960 White to Blue Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10159960 Supplier Macherey-Nagel™ Supplier No. 91312
  • ONLINE EXCLUSIVE PRICE
    403.00 NOK
    380.99 NOK / Pack of 100
    Save 22.01 NOK
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Allows for the quick and easy determination of peroxide levels in solutions. Macherey Nagel™ Quantofix™ Peroxide 100 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Allows for the quick and easy determination of peroxide levels in solutions. Macherey Nagel™ Quantofix™ Peroxide 100 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
  • Ready to use
  • Rapid: provides reliable results within 15 seconds
  • CE marked and suitable to be used in the healthcare environment
  • Can be used in chemical laboratories to check organic solvents
  • Test for residual peroxide in cleaning procedures with high potency disinfectants
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store at room temperature
Specifications
Color Indicator White to Blue
Format Strip
For Use With (Application) Determination of Peroxide 100
Graduations 0, 1, 3, 10, 30, 100mg/L H2O2
Quantity 100 Pack
Storage Requirements Room Temperature
Test Time 15 sec.
Type Semi-qualitative Test Strip