missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Quantofix™ EDTA Semi-quantitative Test Strips

Product Code. 10554598 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color Indicator:
Red to Yellow
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Indicator unitSize
10554598 Red to Yellow Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10554598 Supplier Macherey-Nagel™ Supplier No. 91335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents

  • Rapid: most tests can be carried out in 10s to 120s
  • Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
  • Precise: color charts adjusted and checked using certified standard solutions
  • Shelf life: 2. 5 years, store below 30°C

Specifications

Color Indicator Red to Yellow
Format Strip
For Use With (Application) Determination of EDTA
Graduations 0, 100, 200, 300, 400mg/L EDTA
Quantity 100 Pack
Storage Requirements 30°C
Test Time 15 sec.
Type Semi-qualitative Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.